Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP1/L-FABP Antibody (966927), Alexa Fluor 594, Novus Biologicals™
SDP

Catalog No. MB15651A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB15651A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB15651A594 Supplier Novus Biologicals Supplier No. MAB15651AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP1/L-FABP Monoclonal specifically detects FABP1/L-FABP in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen FABP1/L-FABP
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 966927
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein
Gene Symbols FABP1
Host Species Mouse
Immunogen E. coli-derived recombinant rat FABP1/L-FABP
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2168.0
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.