Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP1/L-FABP Antibody (FABP1/3482), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB378231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378231 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378231 Supplier Novus Biologicals Supplier No. NBP308934AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP1/L-FABP Monoclonal antibody specifically detects FABP1/L-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen FABP1/L-FABP
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone FABP1/3482
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein
Host Species Mouse
Immunogen Human recombinant FABP1/L-FABP protein fragment (around aa1-127) (exact sequence is proprietary) (Uniprot: P07148)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2168
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.