Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP2/I-FABP Antibody (CPTC-FABP2-3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB389117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389117 Supplier Novus Biologicals Supplier No. NBP308950AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen FABP2/I-FABP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-FABP2-3
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Host Species Mouse
Immunogen Recombinant full length human FABP2/I-FABP protein (Uniprot: P12104)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2169
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.