Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP2/I-FABP Antibody, Novus Biologicals™
SDP

Catalog No. NB10059746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10059746 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10059746 Supplier Novus Biologicals Supplier No. NB10059746
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 7 publications

FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Fish, Human, Mouse samples. It is validated for Flow Cytometry, IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen FABP2/I-FABP
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.001 ug/ml, Immunohistochemistry 2 - 3 ug/ml, Immunohistochemistry-Paraffin 2-3 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P12104
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Gene Symbols FABP2
Host Species Goat
Immunogen Peptide with sequence C-EGVEAKRIFKKD corresponding to C-Terminus according to NP_000125.2.
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2169
Test Specificity Publications have used this antibody with Atlantic salmon (Salmo salar) and rainbow trout (Oncorhynchus mykiss). Predicted cross-reactivity based on sequence identity: Rat, Pig, Dog and Cow.
Target Species Fish, Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.