Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP2/I-FABP Antibody (OTI2C4), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N270699APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270699APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270699APC7 Supplier Novus Biologicals Supplier No. NBP270699APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FABP2/I-FABP
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2C4
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Gene Symbols FABP2
Host Species Mouse
Immunogen Full length recombinant protein of human FABP2/I-FABP (NP_000125) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2169.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.