Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP2/I-FABP Rabbit anti-Mouse, Rat, Clone: 5W9L6, Novus Biologicals™
SDP

Catalog No. NB168526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168526 20 μg
NB168525 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168526 Supplier Novus Biologicals Supplier No. NBP31671720UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen FABP2/I-FABP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5W9L6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FABP2/I-FABP (P12104).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2169
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.