Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP3/H-FABP Antibody (9F10), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB162788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162788 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162788 Supplier Novus Biologicals Supplier No. NBP260234C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen FABP3/H-FABP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 9F10
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP
Host Species Mouse
Immunogen Recombinant human FABP3/H-FABP (1-133aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2170
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.