Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP3/H-FABP Antibody (SR1210), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32177950UL
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
FABP3/H-FABP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1210 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
A synthesized peptide derived from human FABP3/H-FABP (Uniprot #: P05413) | |
50 μg | |
Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
2170 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction