Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP3/H-FABP Antibody (SR1210), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
| Antigen | FABP3/H-FABP |
|---|---|
| Clone | SR1210 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| FABP3/H-FABP | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
| A synthesized peptide derived from human FABP3/H-FABP (Uniprot #: P05413) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1210 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2170 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title