Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP4/A-FABP Antibody (009), DyLight 488
SDP

Catalog No. NB069351 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069351 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069351 Supplier Novus Biologicals Supplier No. NBP290055G

Rabbit Monoclonal Antibody

FABP4/A-FABP Monoclonal specifically detects FABP4/A-FABP in Human samples. It is validated for ELISA.

Specifications

Antigen FABP4/A-FABP
Applications ELISA
Classification Monoclonal
Clone 9
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FABP4/A-FABP (P15090; Cys2-Ala132,29Ala/Thr).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2167
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.