Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP5/E-FABP Antibody (311215), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB313111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313111 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313111 Supplier Novus Biologicals Supplier No. FAB3077PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FABP5/E-FABP
Applications Flow Cytometry
Classification Monoclonal
Clone 311215
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog
Host Species Rat
Immunogen E. coli-derived recombinant human FABP5/E-FABP, Ala2-Glu135, Accession # Q01469
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2171
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.