Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP5/E-FABP Antibody (FABP5/3750), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB012793 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012793 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012793 Supplier Novus Biologicals Supplier No. NBP279929FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FABP5/E-FABP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FABP5/3750
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog
Gene Symbols FABP5
Host Species Mouse
Immunogen Recombinant full-length human FABP5/E-FABP protein (Uniprot: Q01469)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.