Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP6 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB322790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322790 Supplier Novus Biologicals Supplier No. NBP300020APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FABP6 Polyclonal antibody specifically detects FABP6 in Human samples. It is validated for ELISA

Specifications

Antigen FABP6
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15PIntestinal 15 kDa protein, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBPIleal lipid-binding protein, illeal lipid-binding protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human FABP6 (Accession#: AAH22489.1; Met 1-Ala 128)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2172
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.