Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP7/B-FABP Antibody (1F5), Novus Biologicals™
SDP

Catalog No. NB119851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119851 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119851 Supplier Novus Biologicals Supplier No. NBP311226
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP7/B-FABP Monoclonal specifically detects FABP7/B-FABP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoassay.

Specifications

Antigen FABP7/B-FABP
Applications Western Blot, Flow Cytometry, Immunoassay
Classification Monoclonal
Clone 1F5
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related
Host Species Mouse
Immunogen Peptide derived from human B-FABP sequence
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2173
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.