Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP7/B-FABP Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB086557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086557 Supplier Novus Biologicals Supplier No. NBP300016AF700

Rabbit Polyclonal Antibody

FABP7/B-FABP Polyclonal specifically detects FABP7/B-FABP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FABP7/B-FABP
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FABP7/B-FABP (O15540; Met1-Ala132).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2173
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.