Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP8/M-FABP/Myelin P2 Protein Antibody (3H3F4D5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350899 Supplier Novus Biologicals Supplier No. NBP306650MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP8/M-FABP/Myelin P2 Protein Monoclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISA

Specifications

Antigen FABP8/M-FABP/Myelin P2 Protein
Applications ELISA
Classification Monoclonal
Clone 3H3F4D5
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PMP2 (Accession#: NP_002668.1; Ser 2-Val 132).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5375
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.