Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP8/M-FABP/Myelin P2 Protein Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB323663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323663 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323663 Supplier Novus Biologicals Supplier No. NBP299794APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FABP8/M-FABP/Myelin P2 Protein Polyclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISA

Specifications

Antigen FABP8/M-FABP/Myelin P2 Protein
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human FABP8/M-FABP/Myelin P2 Protein (Accession#: NP_002668.1; Ser 2-Val 132)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5375
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.