Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP9/T-FABP Antibody (13F9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP23120005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23120005 0.05 mL
NBP23120001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP23120005 Supplier Novus Biologicals Supplier No. NBP2312000.05ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen FABP9/T-FABP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 13F9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000
Formulation PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Gene Accession No. NP_001073995
Gene Alias FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP
Host Species Mouse
Immunogen Recombinant human FABP9 (1-132aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646480
Test Specificity Anti-human FABP9 mAb, clone 13F9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FABP9 protein.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.