Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FACA/FANCA Antibody (SR1233), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | FACA/FANCA |
---|---|
Clone | SR1233 |
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1:40 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
FACA/FANCA Monoclonal antibody specifically detects FACA/FANCA in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
FACA/FANCA | |
Western Blot 1:500-1:1000, Immunoprecipitation 1:40 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FA, FA1, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA | |
A synthesized peptide derived from human FACA/FANCA (Uniprot #: O15360) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1233 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2175 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title