Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FACA/FANCA Antibody (SR1233), Novus Biologicals™
SDP

Catalog No. NB260438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260438 100 μg
NB260447 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260438 Supplier Novus Biologicals Supplier No. NBP322105100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FACA/FANCA Monoclonal antibody specifically detects FACA/FANCA in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen FACA/FANCA
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1233
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 1:40
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias FA, FA1, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA
Host Species Rabbit
Immunogen A synthesized peptide derived from human FACA/FANCA (Uniprot #: O15360)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2175
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.