Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor IX Antibody (3G3), Novus Biologicals™
SDP

Catalog No. NB297791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297791 100 μL
NB297790 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297791 Supplier Novus Biologicals Supplier No. NBP326593100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Factor IX Monoclonal antibody specifically detects Factor IX in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Factor IX
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3G3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Factor IX [UniProt P00740]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2158
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.