Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor IX Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB053133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053133 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053133 Supplier Novus Biologicals Supplier No. NBP298270F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Factor IX Polyclonal specifically detects Factor IX in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Factor IX
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Factor IX (P16294; Met1-Thr471).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2158
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.