Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor V Antibody (2280A), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345604 Supplier Novus Biologicals Supplier No. FAB9856MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Factor V Monoclonal specifically detects Factor V in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry (Intracellular Staining).

Specifications

Antigen Factor V
Applications CyTOF, ELISA, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 2280A
Conjugate mFluor Violet 610 SE
Dilution ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Activated protein C cofactor, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor
Host Species Rabbit
Immunogen Synthetic peptide containing human Coagulation Factor V aa1650-1700, Accession # P12259
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2153
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.