Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Antibody (226), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB334138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334138 Supplier Novus Biologicals Supplier No. NBP290254MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Factor VIII Monoclonal antibody specifically detects Factor VIII in Human samples. It is validated for ELISA

Specifications

Antigen Factor VIII
Applications ELISA
Classification Monoclonal
Clone 226
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Factor VIII
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2157
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.