Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Antibody (2A5), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10062559G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062559G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062559G Supplier Novus Biologicals Supplier No. NB10062559G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor VIII A2 domain Monoclonal specifically detects Factor VIII A2 domain in Human samples. It is validated for ELISA.

Specifications

Antigen Factor VIII A2 domain
Applications ELISA
Classification Monoclonal
Clone 2A5
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Gene Symbols F8
Host Species Mouse
Immunogen Purified human factor VIII
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2157
Test Specificity NB100-62559 recognizes the 83 kDa light chain of Factor VIII. It does not cross-react with Von Willebrand factor.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.