Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$499.50

Specifications

Antigen Factor VIII A2 domain
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP168934
SDP
View Documents
Novus Biologicals
NBP168934
100 μL
Each of 1 for $499.50
Only null left
Add to Cart
 
Description

Description

Factor VIII A2 domain Polyclonal specifically detects Factor VIII A2 domain in Human samples. It is validated for Western Blot.
Specifications

Specifications

Factor VIII A2 domain
Polyclonal
Rabbit
P00451
2157
Synthetic peptides corresponding to F8 (coagulation factor VIII, procoagulant component) The peptide sequence was selected from the C terminal of F8. Peptide sequence IMVTFRNQASRPYSFYSSLISYEEDQRQGAEPRKNFVKPNETKTYFWKVQ.
Primary
Western Blot
Unconjugated
RUO
AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
F8
IgG
79 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.