Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Antibody (RFF-VIII:c/8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395438 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395438 Supplier Novus Biologicals Supplier No. NBP2503980.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor VIII Monoclonal antibody specifically detects Factor VIII in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Functional Assay

Specifications

Antigen Factor VIII
Applications Western Blot, Immunofluorescence, Functional Assay
Classification Monoclonal
Clone RFF-VIII:c/8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Functional
Formulation PBS
Gene Alias AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Host Species Mouse
Immunogen Purified human factor VIII
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2157
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.