Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII:c heavy chain Antibody (RFF-VIII:c/10), PE, Novus Biologicals™
SDP

Catalog No. NB309158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309158 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309158 Supplier Novus Biologicals Supplier No. NBP253119PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor VIII:c heavy chain Monoclonal antibody specifically detects Factor VIII:c heavy chain in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Factor VIII:c heavy chain
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone RFF-VIII:c/10
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Host Species Mouse
Immunogen Purified human factor VIII
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.