Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII heavy chain Antibody (10-11-37), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB241377700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB241377700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB241377700 Supplier Novus Biologicals Supplier No. NBP241377AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.

Specifications

Antigen Factor XII heavy chain
Applications Western Blot, ELISA
Classification Monoclonal
Clone 13799
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor
Gene Symbols F12
Host Species Mouse
Immunogen Affinity purified human factor XII.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2161
Test Specificity Antibody binds the heavy chain of human factor XII, including factor XII derivatives such as a-factor XIIa and a-factor XIIa C1-inhibitor complexes.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.