Learn More
Factor XII heavy chain Antibody (10-11-37), Biotin, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Factor XII heavy chain |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 13799 |
| Conjugate | Biotin |
| Dilution | Western Blot, ELISA, Sandwich ELISA |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor |
| Gene Symbols | F12 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.