Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII heavy chain Antibody (10-11-5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP24137A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24137A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24137A30 Supplier Novus Biologicals Supplier No. NBP241375AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.

Specifications

Antigen Factor XII heavy chain
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10-11-5
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor
Gene Symbols F12
Host Species Mouse
Immunogen Affinity purified human factor XII.
Purification Method Protein A or G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2161.0
Test Specificity Antibody binds the heavy chain of human factor XII, including factor XII derivatives such as a-factor XIIa and a-factor XIIa C1-inhibitor complexes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.