Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII heavy chain Antibody (OTI1H5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270695V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270695V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270695V Supplier Novus Biologicals Supplier No. NBP270695V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Factor XII heavy chain
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1H5
Conjugate DyLight 405
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor
Gene Symbols F12
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 73-372 of human Factor XII heavy chain (NP_000496) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2161.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.