Learn More
Description
Specifications
Specifications
| Antigen | Factor XIIIa |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AC-1A1 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 |
| Formulation | EDTA, pH 8.0, containing less than 0.1% sodium azide with Sodium Azide |
| Gene Accession No. | P00488 |
| Gene Alias | bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
