Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIa Antibody (AC-1A1), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $853.50
Specifications
| Antigen | Factor XIIIa |
|---|---|
| Clone | AC-1A1 |
| Concentration | 0.1 mg/mL |
| Dilution | Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
Description
Factor XIIIa Monoclonal specifically detects Factor XIIIa in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Factor XIIIa | |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology | |
| P00488 | |
| 2162 | |
| BALB/C mice were injected with recombinant human protein corresponding to A-subunit of coagulation Factor XIII. | |
| Primary | |
| Store at 4°C. |
| AC-1A1 | |
| Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma | |
| F13A1 | |
| IgG1 κ | |
| Protein A or G purified | |
| This is specific to 160 kDa protein known as Factor XIIIA. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title