Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XIIIa Antibody (F13A1/1448), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP259605AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259605AI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259605AI Supplier Novus Biologicals Supplier No. NBP259605AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor XIIIa Monoclonal specifically detects Factor XIIIa in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Factor XIIIa
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone F13A1/1448
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma
Gene Symbols F13A1
Host Species Mouse
Immunogen Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) (Uniprot: P00488)
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 2162
Test Specificity The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. It recognizes a protein of 83kDa, which is identified as Factor XIIIa. It has been identified in platelets, megakaryocytes, and fibroblast-like mesenchymal or histiocytic cells in the placenta, uterus, and prostate, monocytes and macrophages and dermal dendritic cells. Anti-factor XIIIa has been found to be useful in differentiating between dermatofibroma (almost all cases are positive), dermatofibrosarcoma protuberans (-/+) and desmoplastic malignant melanoma (-). Anti-factor XIIIa positivity is also seen in capillary hemagioblastoma, hemangioendothelioma, hemangiopericytoma, xanthogranuloma, xanthoma, hepatocellular carcinoma, glomus tumor, and meningioma.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.