Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIa Mouse anti-Human, Clone: F13A1/1448, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $526.00
Specifications
Antigen | Factor XIIIa |
---|---|
Clone | F13A1/1448 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5-1.0 ug/ml, Flow Cytometry 0.5-1 ug/million cells, ELISA 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page, Protein Array 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25960420
![]() |
Novus Biologicals
NBP25960420UG |
20 μg |
Each for $268.00
|
|
|||||
NBP259604X
![]() |
Novus Biologicals
NBP259604100UG |
100 μg |
Each for $526.00
|
|
|||||
Description
Factor XIIIa Monoclonal specifically detects Factor XIIIa in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
Factor XIIIa | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Cell Biology | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
2162 | |
Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
83 kDa |
F13A1/1448 | |
Western Blot 0.5-1.0 ug/ml, Flow Cytometry 0.5-1 ug/million cells, ELISA 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page, Protein Array 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma | |
F13A1 | |
IgG2b κ | |
Protein A or G purified | |
The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. It recognizes a protein of 83kDa, which is identified as Factor XIIIa. It has been identified in platelets, megakaryocytes, and fibroblast-like mesenchymal or histiocytic cells in the placenta, uterus, and prostate, monocytes and macrophages and dermal dendritic cells. Anti-factor XIIIa has been found to be useful in differentiating between dermatofibroma (almost all cases are positive), dermatofibrosarcoma protuberans ( -/+) and desmoplastic malignant melanoma (-). Anti-factor XIIIa positivity is also seen in capillary hemagioblastoma, hemangioendothelioma, hemangiopericytoma, xanthogranuloma, xanthoma, hepatocellular carcinoma, glomus tumor, and meningioma. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title