Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FADD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP14555225UG
Description
FADD Polyclonal specifically detects FADD in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| FADD | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 5 mM EDTA, 50% glycerol, 0.5% BSA with 0.03% Proclin 300 | |
| Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
| Rabbit | |
| 23 kDa | |
| 25 μg | |
| Apoptosis, Cancer, Diabetes Research, Tumor Suppressors | |
| 8772 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 4 μg/mL, Immunoprecipitation | |
| Q13158 | |
| FADD | |
| Synthetic peptide surrounding amino acid 128 of human FADD. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction