Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FADD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$201.50
Specifications
| Antigen | FADD |
|---|---|
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FADD Polyclonal specifically detects FADD in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| FADD | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Diabetes Research, Tumor Suppressors | |
| PBS (pH 7.2), 5 mM EDTA, 50% glycerol, 0.5% BSA with 0.03% Proclin 300 | |
| Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
| FADD | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate, Monkey | |
| Q13158 | |
| 8772 | |
| Synthetic peptide surrounding amino acid 128 of human FADD. | |
| Primary | |
| 23 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title