Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Antibody (OTI1C11), PE, Novus Biologicals™
SDP

Catalog No. NBP271387PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271387PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271387PE Supplier Novus Biologicals Supplier No. NBP271387PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FADD Monoclonal specifically detects FADD in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FADD
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C11
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD
Gene Symbols FADD
Host Species Mouse
Immunogen Full length recombinant protein of human FADD (NP_003815) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8772.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.