Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Rabbit anti-Human, Mouse, Rat, Clone: 3X9R6, Novus Biologicals™
SDP

Catalog No. NB166997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166997 100 μg
NB166998 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166997 Supplier Novus Biologicals Supplier No. NBP315951100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FADD Monoclonal antibody specifically detects FADD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FADD
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3X9R6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of mouse FADD (NP_034305.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8772
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.