Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAHD2A Antibody (OTI6D9), FITC, Novus Biologicals™
SDP

Catalog No. NBP271886F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271886F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271886F Supplier Novus Biologicals Supplier No. NBP271886F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAHD2A Monoclonal specifically detects FAHD2A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAHD2A
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6D9
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias CGI-105, EC 3.-, fumarylacetoacetate hydrolase domain containing 1, fumarylacetoacetate hydrolase domain containing 2A, fumarylacetoacetate hydrolase domain-containing protein 2A, MGC131995
Gene Symbols FAHD2A
Host Species Mouse
Immunogen Full length recombinant protein of human FAHD2A (NP_057128) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51011.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.