Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAHD2B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | FAHD2B |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FAHD2B Polyclonal specifically detects FAHD2B in Mouse samples. It is validated for Western Blot.Specifications
FAHD2B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
DKFZp434N062, EC 3.-, fumarylacetoacetate hydrolase domain containing 2B, fumarylacetoacetate hydrolase domain-containing protein 2B | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse FAHD2B (NP_083905). Peptide sequence VSQFVTLYPGDLLLTGTPPGVGMFRKPPVFLKKGDEVQCEIEELGVIINK | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
151313 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title