Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAIM2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224713 0.1 mg
NBP224713SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224713 Supplier Novus Biologicals Supplier No. NBP224713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

FAIM2 Polyclonal specifically detects FAIM2 in Canine, Human, Mouse, Primate, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen FAIM2
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 26565411), Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9BWQ8
Gene Alias Fas apoptotic inhibitory molecule 2, KIAA0950NGP35, LFG2, LFGLIFEGUARD, NMP35neural membrane protein 35, Protein lifeguard, TMBIM2lifeguard, transmembrane BAX inhibitor motif containing 2, Transmembrane BAX inhibitor motif-containing protein 2
Gene Symbols FAIM2
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human Lifeguard was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 23017
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a, CRA_b, CRA_c and CRA_d).
Target Species Canine, Human, Mouse, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.