Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAIM2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen FAIM2
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 26565411), Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224713SS
SDP
View Documents
Novus Biologicals
NBP224713SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224713
SDP
View Documents
Novus Biologicals
NBP224713
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

FAIM2 Polyclonal specifically detects FAIM2 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FAIM2
Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 26565411), Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat, Canine, Primate
Q9BWQ8
23017
A portion of amino acids 1-50 of human Lifeguard was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis
PBS with 0.05% Sodium Azide
Fas apoptotic inhibitory molecule 2, KIAA0950NGP35, LFG2, LFGLIFEGUARD, NMP35neural membrane protein 35, Protein lifeguard, TMBIM2lifeguard, transmembrane BAX inhibitor motif containing 2, Transmembrane BAX inhibitor motif-containing protein 2
FAIM2
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a, CRA_b, CRA_c and CRA_d).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.