Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAIM3 Antibody (992338), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345703 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345703 Supplier Novus Biologicals Supplier No. FAB9494MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAIM3 Monoclonal specifically detects FAIM3 in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen FAIM3
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 992338
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, TOSOIgM Fc receptor
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession # O60667
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9214
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.