Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FALDH Antibody (OTI2A7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP270159IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270159IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270159IR Supplier Novus Biologicals Supplier No. NBP270159IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FALDH Monoclonal specifically detects FALDH in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen FALDH
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2A7
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS
Gene Symbols ALDH3A2
Host Species Mouse
Immunogen Full length recombinant protein of human FALDH (NP_001026976) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 224.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.