Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM101A Antibody, Novus Biologicals™
SDP

Catalog No. NB393419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393419 25 μL
NBP258508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393419 Supplier Novus Biologicals Supplier No. NBP25850825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM101A Polyclonal specifically detects FAM101A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAM101A
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias family with sequence similarity 101, member A, FLJ44614
Gene Symbols FAM101A
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FFGESIKVNPEPTHEIRCNSEVKYASEKHFQDKVFYAPVPTITAYSETIVAAPNCTWRNYRSQLTLEPRPRALRFRSTTIIFPKHARSTFRTTLHCSLGRPSRWFTASVQLQLCQDP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144347
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.