Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM105A Antibody, Novus Biologicals™
SDP

Catalog No. NBP15983520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15983520 20 μL
NBP159835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15983520 Supplier Novus Biologicals Supplier No. NBP15983520UL

Rabbit Polyclonal Antibody

FAM105A Polyclonal specifically detects FAM105A in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM105A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NUU6
Gene Alias family with sequence similarity 105, member A, FLJ11127, FLJ43660, hypothetical protein LOC54491, NET20
Gene Symbols FAM105A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FAM105A (family with sequence similarity 105, member A) The peptide sequence was selected from the N terminal of FAM105A. Peptide sequence HKLKWWIGYLQRKFKRNLSVEAEVDLLSYCAREWKGETPRNKLMRKAYEE.
Molecular Weight of Antigen 42 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54491
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.