Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM107A Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB044871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044871 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044871 Supplier Novus Biologicals Supplier No. NBP297556JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM107A Polyclonal specifically detects FAM107A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FAM107A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias downregulated in renal cell carcinoma, DRR1family with sequence similarity 107 member A transcript, family with sequence similarity 107, member A, FLJ30158, FLJ45473, Protein TU3A, TU3ADown-regulated in renal cell carcinoma 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM107A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11170
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.