Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM119A Antibody (OTI2G7), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP272423UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272423UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272423UV Supplier Novus Biologicals Supplier No. NBP272423UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM119A Monoclonal specifically detects FAM119A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen FAM119A
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2G7
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.1.1.-, FAM119A, HCA557B, Hepatocellular carcinoma-associated antigen 557b, hypothetical protein LOC151194, LOC151194, member A, methyltransferase like 21A, MGC45373
Gene Symbols METTL21A
Host Species Mouse
Immunogen Full length recombinant protein of human FAM119A (NP_660323) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 151194.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.